Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-m-toluidine 97%

Supplier: Ambeed
Danger

A134641-1G A134641-10G A134641-25G A134641-5G
77190-148EA 32 USD InStock
77190-148 77190-150 77190-146 77190-144
6-Chloro-m-toluidine 97%
6-Chloro-m-toluidine
Caution: for Research Use Only


Formula: C₇H₈ClN
MW: 141.6 g/mol
MDL Number: MFCD00007674
CAS Number: 95-81-8
UN: 2239
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty