Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylmalonic acid 97%

Supplier: Ambeed
Warning

A143479-10G A143479-25G A143479-1G A143479-5G
77162-444EA 27 USD InStock
77162-444 77162-446 77162-448 77162-450
Ethylmalonic acid 97%
Ethylmalonic acid
Caution: for Research Use Only


Formula: C₅H₈O₄
MDL Number: MFCD00002668
CAS Number: 601-75-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty