Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Thienyl)-L-alanine 97%

Supplier: Ambeed
Warning
Synonyms: H-β-(2-Thienyl)-Ala-OH(S)-2-Amino-3-(thiophen-2-yl)propanoic acid

A178080-1G A178080-250MG
77191-610EA 32 USD InStock
77191-610 77191-612
3-(2-Thienyl)-L-alanine 97%
3-(2-Thienyl)-L-alanine
Caution: for Research Use Only


Formula: C₇H₉NO₂S
MW: 171.214 g/mol
MDL Number: MFCD00064335
CAS Number: 22951-96-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty