Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Anisohydrazide 98%

Supplier: Ambeed
Warning
Synonyms: 2-Methoxybenzene-1-carbohydrazide2-Anisohydrazide2-Methoxybenzoic acid hydrazide2-Methoxybenzoic hydrazide2-Methoxybenzohydrazide2-Methoxybenzhydrazide

A211458-100G A211458-5G
77192-660EA 9 USD InStock
77192-660 77192-658
o-Anisohydrazide 98%
o-Anisohydrazide
Caution: for Research Use Only


Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00014755
CAS Number: 7466-54-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty