Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Triethynylbenzene 98%

Supplier: Ambeed
Warning

A480200-5G A480200-250MG A480200-1G A480200-25G
77196-778EA 63 USD InStock
77196-778 77196-772 77196-774 77196-776
1,3,5-Triethynylbenzene 98%
1,3,5-Triethynylbenzene
Caution: for Research Use Only


Formula: C₁₂H₆
MW: 150.18 g/mol
MDL Number: MFCD00068621
CAS Number: 7567-63-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty