Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) 98%

Supplier: Ambeed
Warning
Synonyms: 1,4,7,10-Tetraoxa-13-azacyclopentadecane

A505766-250MG A505766-1G
77197-076EA 50 USD InStock
77197-076 77197-074
1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) 98%
1-Aza-15-crown-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecane)
Caution: for Research Use Only


Formula: C₁₀H₂₁NO₄
MW: 219.28 g/mol
MDL Number: MFCD00075465
CAS Number: 66943-05-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty