Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-5-methylbenzene-1,2-diamine 97%

Supplier: Ambeed
Warning
Synonyms: 3-Bromo-4,5-diaminotoluene3-Bromo-5-methylphenylene-1,2-diamine

A558240-250MG
77169-800EA 59 USD InStock
77169-800
3-Bromo-5-methylbenzene-1,2-diamine 97%
3-Bromo-5-methylbenzene-1,2-diamine
Caution: for Research Use Only


Formula: C₇H₉BrN₂
MW: 201.064 g/mol
MDL Number: MFCD18088524
CAS Number: 70733-25-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty