Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Propanedisulfonic acid disodium salt 98%

Supplier: Ambeed
Warning
Synonyms: Sodium propane-1,3-disulphonateSodium propane-1,3-disulfonate

A564757-100MG
77197-672EA 5 USD InStock
77197-672
1,3-Propanedisulfonic acid disodium salt 98%
1,3-Propanedisulphonic acid disodium salt
Caution: for Research Use Only


Formula: C₃H₆Na₂O₆S₂
MW: 248.19 g/mol
MDL Number: MFCD00191483
CAS Number: 36589-58-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty