Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,3,3-Tetramethoxypropane 97%

Supplier: Ambeed
Danger
Synonyms: Malonaldehyde bis(dimethyl acetal)

A647000-25G A647000-10G
77198-412EA 5 USD InStock
77198-412 77198-414
1,1,3,3-Tetramethoxypropane 97%
1,1,3,3-Tetramethoxypropane
Caution: for Research Use Only


Formula: C₇H₁₆O₄
MW: 164.2 g/mol
MDL Number: MFCD00008488
CAS Number: 102-52-3
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty