Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Pantolactone 98%

Supplier: Ambeed
Warning
Synonyms: (R)-(-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanonePantolactoneD-(-)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone(R)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-oneD-(-)-Pantoyl Lactone

A762508-5G A762508-500G A762508-10G A762508-100G A762508-25G
77171-140EA 6 USD InStock
77171-140 77171-142 77171-144 77171-146 77171-138
D-(-)-Pantolactone 98%
D-(-)-Pantolactone
Caution: for Research Use Only


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00005392
CAS Number: 599-04-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty