Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Amino-1,2-propanediol 95%

Supplier: Ambeed
Danger
Synonyms: (±)-3-Amino-1,2-dihydroxy propane3-Amino-1,2-propanediol

A853353-25G
77171-654EA 6 USD InStock
77171-654
(±)-3-Amino-1,2-propanediol 95%
(±)-3-Amino-1,2-propanediol
Caution: for Research Use Only


Formula: C₃H₉NO₂
MW: 91.11 g/mol
MDL Number: MFCD00008140
CAS Number: 616-30-8
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty