Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-D-Galactose pentaacetate 95%

Supplier: Ambeed
Warning
Synonyms: a-D-Galactose pentaacetate

A870142-100G A870142-25G A870142-5G
77200-080EA 13 USD InStock
77200-080 77200-078 77200-076
α-D-Galactose pentaacetate 95%
alpha-D-Galactose pentaacetate
Caution: for Research Use Only


Formula: C₁₆H₂₂O₁₁
MW: 390.341 g/mol
MDL Number: MFCD00064081
CAS Number: 4163-59-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty