Would you like to log in again?
PIP® Arc Shield with Hard Hat
Supplier: Protective Industrial Products
PIP®
9150-56510
76945-074EA 126.99 USD InStock
76945-074
PIP® Arc Shield with Hard Hat
Face Shields
Designed for use anywhere protection is needed from the hazards of arc flash.
  • Arc rating: ATPV 12.1 cal/cm²
  • PPE category: 2
  • Hard hat: 280-EV6151-OR
  • Arc shield: 9400-55505
Ideally suited for use in electrical, utility, industrial, and construction applications.
Certifications: TAA compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty