Would you like to log in again?
NEBNext® UltraShear™ DNA Library Preparation Kits
Supplier: New England Biolabs
NEBNext® UltraShear™
M7634L M7634S
77025-078EA 432 USD InStock
77025-078 77025-088
NEBNext® UltraShear™ DNA Library Preparation Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEBNext UltraShear fragments DNA in a time-dependent manner and the same protocol is followed for all input amounts (5 to 250 ng).
  • Does not affect methylation marks
  • Methylation analysis: improves CpG coverage and sequencing metrics
  • FFPE DNA: improves usable reads and decreases artificial mutation frequency
  • Single protocol for all input amounts
  • Increased library yields
Enzymatic fragmentation of DNA as part of the library prep workflow provides many advantages compared to mechanical shearing. However, specialized fragmentation reagents are required for enzymatic shearing of challenging samples such as FFPE DNA, and in order to maintain methylation marks on samples for methylome analysis. NEBNext UltraShear is a mix of enzymes that has been designed and optimized to fragment these sample types upstream of library preparation. This improves library yields and diversity, and allows retention of methylation marks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty