Would you like to log in again?
Ph.D.-C7C™ Phage Display Peptide Library Kit v2
Supplier: New England Biolabs
Ph.D.-C7C™
E8212S
77025-074EA 786 USD InStock
77025-074
Ph.D.-C7C™ Phage Display Peptide Library Kit v2
Vectors, Plasmids and Libraries
The Ph.D.-C7C Phage display peptide library kit v2 is based on a combinatorial library of random disulfide looped peptides fused to the N-terminus of a minor coat protein (pIII) of M13 phage.
  • Ready to use complex phage library (~1E09 clones)
  • Structurally constrained peptide library
  • Panning against Mouse mAb yields more reliable epitope results than former target protein
  • Does not require helper phage for amplification
  • Inherent link between phenotype and genotype allows screening of billions of clones in a single tube
The library is displayed in the form AC-X7-C-GGG, which directly precedes the wild-type pIII sequence. Under neutral conditions disulfide loop formation is favored. The library consists of ~1E09 electroporated sequences amplified once to yield approximately 100 copies of each sequence in 10 μl of the supplied phage. The Ph.D.-C7C Phage display peptide library kit v2 contains the Ph.D.-C7C Phage display peptide library, a DYKDDDDK Mouse monoclonal antibody and Protein G Magnetic beads for a panning control experiment, and enough -96 g III sequencing primer for >50 sequencing reactions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty