Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGAV Mouse Monoclonal Antibody [clone: [13C2]] (PE (Phycoerythrin))

Supplier: ProSci
99-552
77119-278EA 412.78 USD InStock
77119-278
Anti-ITGAV Mouse Monoclonal Antibody [clone: [13C2]] (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: ITGAV
Clonality: Monoclonal
Clone: [13C2]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, African Green Monkey, Rabbit, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty