Would you like to log in again?
AccuTec Pro Utility Blades, 2-Notch
Supplier: AccuTec Blades
AccuTec Pro
APBL-8018-0000
77172-144CS 415.67 USD InStock
77172-144
AccuTec Pro Utility Blades, 2-Notch
Razors and Blades
AccuTec Pro utility blades feature two-facet edges for durability. Multi-purpose blades are constructed with heavy-duty, high carbon steel and fit most standard utility knives.
  • Two-facet edge for durability
  • Individually wrapped for safety
  • Each carton contains 100 blades
Blades fit most standard utility knives.
Case dimensions: 9.0×5.5×3.25"
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty