Would you like to log in again?
AccuTec Pro Hobby Knife and Blades
Supplier: AccuTec Blades
AccuTec Pro
APTL-5000-0000
77172-146CS 586 USD InStock
77172-146
AccuTec Pro Hobby Knife and Blades
Knives
AccuTec Pro No.1 hobby knife and #11 hobby blades are great for detailed cutting and trimming of lightweight materials.
  • Sure-grip barrel for comfort and control
  • Four jaw chuck
  • Heavy-duty knurled collar for durability
  • Plastic safety cap protects knife when not in use
This set includes five #11 blades in a storage tube. The hobby knife is compatible with #10, #11, #16, and #17 hobby blades.
Case dimensions: 10.44×7.88×9.75"
Packaging: Each card contains one hobby knife and five #11 hobby blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty