Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco Wicks for Alcohol Lamps, Pack of 10

Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0533WICKPK10
470344-232EA 12.5 USD InStock
470344-232
Eisco Wicks for Alcohol Lamps, Pack of 10
Heaters Spirit Lamps
Replacement set of 100% round cotton wicks for alcohol lamps.

  • Set of 10 round cotton wicks made of 100% cotton
  • Use with alcohol lamps, oil lamps, citronella lamps, and paraffin oil lamps
  • Also good for use with plants and hydroponics as an automatic watering device system
  • Wicks are unprimed and lead-free
  • Each wick measures 0.15" in diameter

Fuel soaks up the wick and produces a flame when the exposed end is lit with a match or lighter. A change of fuel type requires a change of wick. Used to produce an open flame. When using with an alcohol lamp, wicks produce a cooler flame than a propane or butane burner. For alcohol lamps, use with 95% denatured ethanol.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty