Would you like to log in again?
Wash Plate
Supplier: Zymo Research
C2024 C2024-1
77020-272EA 21.95 USD InStock
77020-272 77020-276
Wash Plate
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Wash Plate can be used with centrifuges and vacuum manifolds to minimize cross-contamination between wells.
  • The capacity of the well is 1.3 ml
Available as a refill for the ZymoPURE™ 96 Plasmid Miniprep Kit. The Wash Plate features durable polypropylene construction and can be used with centrifuges and vacuum manifolds to minimize cross-contamination between wells when used in conjunction with our Zymo-Spin P-96 Plate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty