Would you like to log in again?
Vernier Go Direct® Projectile Launcher
Supplier: VERNIER SOFTWARE & TECH SE
GDX-PL
470311-382EA 498.99 USD InStock
470311-382
Vernier Go Direct® Projectile Launcher
Educational Materials Physics Educational Materials Mechanics Learning Activities
Reliable, quantitative data over Bluetooth to investigate important concepts in projectile motion.
  • Extremely precise and reliable
  • Bluetooth device and free software are compatible with PC, Mac, and Chromebooks
  • Pneumatic launching system
  • Heavy base
  • Built-in photogates
Use the Go Direct Projectile Launcher to investigate important concepts in two-dimensional kinematics. Launch steel balls at angles between 0 and 90 degrees and over distances up to 2.5 m.
The solid, heavy base provides an easy-to-use and reliable setup. A unique pneumatic launching system provides excellent repeatability and allows you to set the launch speed.
Built-in photogates provide easy and accurate measurement of the ball’s launch speed and built-in accelerometers report launch angle, allowing for precise quantitative analysis of projectile motion.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary.
The unit is powered via the included USB cable connected to an available USB port on your device, USB wall outlet, or USB power brick.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty