Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Pyrrolidinyl)pyridine 97%

Supplier: Ambeed
Warning
Synonyms: 4-Pyrrolidin-2-ylpyridine

A104385-1G A104385-250MG
77245-310EA 77 USD InStock
77245-310 77245-312
4-(2-Pyrrolidinyl)pyridine 97%
4-(2-Pyrrolidinyl)pyridine
Caution: for Research Use Only


Formula: C₉H₁₂N₂
MW: 148.21 g/mol
MDL Number: MFCD01862536
CAS Number: 128562-25-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty