Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium-4-aminobenzoate 95%

Supplier: Ambeed
Warning
Synonyms: 4-Aminobenzoic Acid Sodium SaltSodium 4-aminobenzoate

A106394-25G A106394-5G A106394-100G
77283-472EA 14 USD InStock
77283-472 77283-470 77283-474
Sodium-4-aminobenzoate 95%
Sodium-4-aminobenzoate
Caution: for Research Use Only


Formula: C₇H₇NO₂
MW: 159.12 g/mol
MDL Number: MFCD00064395
CAS Number: 555-06-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty