Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromononane 98%

Supplier: Ambeed
Warning
Synonyms: 1-Nonyl bromide

A125696-10G A125696-5G
77234-308EA 6 USD InStock
77234-308 77234-310
1-Bromononane 98%
1-Bromononane
Caution: for Research Use Only


Formula: BrH₂C(CH₂)₇CH₃
MW: 207.15 g/mol
MDL Number: MFCD00000278
CAS Number: 693-58-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty