Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bexarotene 98%

Supplier: Ambeed
Warning
Synonyms: 4-(1-(3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)vinyl)benzoic acid

A129004-10MG A129004-50MG A129004-100MG A129004-1MG A129004-1G
77285-216EA 29 USD InStock
77285-216 77285-208 77285-210 77285-214 77285-212
Bexarotene 98%
Bexarotene
Caution: for Research Use Only


Formula: C₂₄H₂₈O₂
MW: 348.486 g/mol
MDL Number: MFCD00932428
CAS Number: 153559-49-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty