Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dichlorobenzylamine 98%

Supplier: Ambeed
Danger
Synonyms: (2,4-Dichlorophenyl)methanamine

A135698-25G A135698-10G A135698-1G
77246-728EA 58 USD InStock
77246-728 77246-726 77246-724
2,4-Dichlorobenzylamine 98%
2,4-Dichlorobenzylamine
Caution: for Research Use Only


Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
MDL Number: MFCD00008109
CAS Number: 95-00-1
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty