Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-β-D-Arabinofuranosyluracil 98%

Supplier: Ambeed
Warning
Synonyms: Uracil 1-β-D-ArabinofuranosideAra-U1-((2R,3S,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

A141122-1G
77246-942EA 6 USD InStock
77246-942
1-β-D-Arabinofuranosyluracil 98%
1-beta-D-Arabinofuranosyluracil
Caution: for Research Use Only


Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
MDL Number: MFCD00065998
CAS Number: 3083-77-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty