Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-3-iodopyridin-4-amine 98%

Supplier: Ambeed
Warning
Synonyms: 4-Amino-2-chloro-3-iodopyridine

A146613-5G A146613-1G
77259-040EA 18 USD InStock
77259-040 77259-042
2-Chloro-3-iodopyridin-4-amine 98%
2-Chloro-3-iodopyridin-4-amine
Caution: for Research Use Only


Formula: C₅H₄ClIN₂
MW: 254.455 g/mol
MDL Number: MFCD09763660
CAS Number: 909036-46-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty