Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-6-guanidinohexanoic acid hydrochloride 95%

Supplier: Ambeed
Warning
Synonyms: H-HomoArg-OH.HClH-HoArg-OH·HClL(+)-Homoarginine hydrochlorideH-HomoArg-OH·HCl

A152121-10G A152121-25MG A152121-10MG
77273-018EA 27 USD InStock
77273-018 77273-020 77273-022
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride 95%
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride
Caution: for Research Use Only


Formula: C₇H₁₆N₄O₂·HCl
MW: 224.7 g/mol
MDL Number: MFCD00012622
CAS Number: 1483-01-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty