Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminonicotinic acid 98%

Supplier: Ambeed
Warning
Synonyms: 5-Amino-3-pyridinecarboxylic acid

A167000-1G A167000-10G A167000-5G
77273-570EA 18 USD InStock
77273-570 77273-574 77273-572
5-Aminonicotinic acid 98%
5-Aminonicotinic acid
Caution: for Research Use Only


Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
MDL Number: MFCD00129116
CAS Number: 24242-19-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty