Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(1H-1,2,4-Triazol-1-yl)benzoic acid 95%

Supplier: Ambeed
Warning
Synonyms: 4-[1,2,4]Triazol-1-yl-benzoic acid

A174379-250MG A174379-1G A174379-5G
77286-954EA 48 USD InStock
77286-954 77286-956 77286-958
4-(1H-1,2,4-Triazol-1-yl)benzoic acid 95%
4-(1H-1,2,4-Triazol-1-yl)benzoic acid
Caution: for Research Use Only


Formula: C₉H₇N₃O₂
MW: 189.1708 g/mol
MDL Number: MFCD02682060
CAS Number: 162848-16-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty