Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorobenzaldehyde 96% (by GC)

Supplier: Ambeed
Danger

A211523-25G
77275-166EA 6 USD InStock
77275-166
3-Fluorobenzaldehyde 96% (by GC)
3-Fluorobenzaldehyde
Caution: for Research Use Only


Formula: C₇H₅FO
MW: 124.11 g/mol
MDL Number: MFCD00003348
CAS Number: 456-48-4
UN: 1989
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty