Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anthranilamide 98%

Supplier: Ambeed
Warning
Synonyms: 2-Aminobenzamide

A226696-100G A226696-10G A226696-25G A226696-500G
77261-438EA 5 USD InStock
77261-438 77303-776 77261-440 77261-436
Anthranilamide 98%
Anthranilamide
Caution: for Research Use Only


Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD00007981
CAS Number: 88-68-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty