Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-chlorobenzene 97%

Supplier: Ambeed
Warning
Synonyms: 4-Bromochlorobenzene

A230755-1000G A230755-25G A230755-500G A230755-100G
77288-374EA 6 USD InStock
77288-374 77288-376 77288-370 77288-372
1-Bromo-4-chlorobenzene 97%
1-Bromo-4-chlorobenzene
Caution: for Research Use Only


Formula: C₆H₄BrCl
MW: 191.45 g/mol
MDL Number: MFCD00000600
CAS Number: 106-39-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty