Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Benzyloxybenzyl alcohol 95%

Supplier: Ambeed
Warning
Synonyms: (4-(Benzyloxy)phenyl)methanol

A236599-100G A236599-25G
77288-606EA 136 USD InStock
77288-606 77288-608
4-Benzyloxybenzyl alcohol 95%
4-Benzyloxybenzyl alcohol
Caution: for Research Use Only


Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD00004654
CAS Number: 836-43-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty