Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-2-pentenoic acid 99%

Supplier: Ambeed
Danger

A248702-10G A248702-5G A248702-25G
77276-264EA 7 USD InStock
77276-264 77276-266 77276-268
2-Methyl-2-pentenoic acid 99%
2-Methyl-2-pentenoic acid
Caution: for Research Use Only


Formula: C₆H₁₀O₂
MDL Number: MFCD00002655
CAS Number: 3142-72-1
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty