Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Pyridin-3-yl-1H-benzimidazole 95%

Supplier: Ambeed
Warning
Synonyms: 2-(Pyridin-3-yl)-1H-benzo[d]imidazole

A249856-10G A249856-1G A249856-5G
77238-252EA 110 USD InStock
77238-252 77238-250 77238-248
2-Pyridin-3-yl-1H-benzimidazole 95%
2-Pyridin-3-yl-1H-benzimidazole
Caution: for Research Use Only


Formula: C₁₂H₉N₃
MW: 195.23 g/mol
MDL Number: MFCD00022665
CAS Number: 1137-67-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty