Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride 98%

Supplier: Ambeed
Warning
Synonyms: (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride

A288973-5G
77289-764EA 174 USD InStock
77289-764
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride 98%
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride
Caution: for Research Use Only


Formula: C₁₃H₂₀BNO₂·HCl
MW: 269.58 g/mol
MDL Number: MFCD02179455
CAS Number: 850568-55-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty