Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Lutidine 98%

Supplier: Ambeed
Danger
Synonyms: 2,3-Dimethylpyridine

A289271-5G A289271-10G A289271-25G
77250-634EA 8 USD InStock
77250-634 77250-632 77250-636
2,3-Lutidine 98%
2,3-Lutidine
Caution: for Research Use Only


Formula: C₇H₉N
MW: 107.16 g/mol
MDL Number: MFCD00009605
CAS Number: 583-61-9
UN: 2924
ADR: 3,8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty