Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentylaniline 98%

Supplier: Ambeed
Warning

A303635-5G A303635-1G A303635-25G
77290-040EA 45 USD InStock
77290-040 77290-036 77290-038
4-Pentylaniline 98%
4-Pentylaniline
Caution: for Research Use Only


Formula: C₁₁H₁₇N
MW: 163.26 g/mol
MDL Number: MFCD00007926
CAS Number: 33228-44-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty