Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-3-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A321911-5G A321911-10G A321911-25G A321911-1G
77239-602EA 5 USD InStock
77239-602 77239-600 77239-606 77239-604
4-Methyl-3-nitrobenzaldehyde 98%
4-Methyl-3-nitrobenzaldehyde
Caution: for Research Use Only


Formula: C₈H₇NO₃
MDL Number: MFCD00017011
CAS Number: 31680-07-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty