Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-2-Chloro-β-nitrostyrene 98% (only trans isomers)

Supplier: Ambeed
Danger
Synonyms: 1-(2-Chlorophenyl)-2-nitroethylene2-Chloro-β-nitrostyrene

A326080-5G A326080-25G A326080-1G
77239-670EA 103 USD InStock
77239-670 77239-672 77239-668
trans-2-Chloro-β-nitrostyrene 98% (only trans isomers)
trans-2-Chloro-β-nitrostyrene
Caution: for Research Use Only


Formula: C₈H₆ClNO₂
MW: 183.60 g/mol
MDL Number: MFCD00024820
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty