Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-chloro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

A329315-25G A329315-1G A329315-100G
77239-740EA 5 USD InStock
77239-740 77239-742 77239-738
1-Bromo-2-chloro-4-nitrobenzene 98%
1-Bromo-2-chloro-4-nitrobenzene
Caution: for Research Use Only


Formula: C₆H₃BrClNO₂
MDL Number: MFCD00051514
CAS Number: 29682-39-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty