Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-nitrothiazole 98%

Supplier: Ambeed
Warning

A342770-25G A342770-10G A342770-1G A342770-5G
77290-552EA 64 USD InStock
77290-552 77290-554 77290-556 77290-558
2-Bromo-5-nitrothiazole 98%
2-Bromo-5-nitrothiazole
Caution: for Research Use Only


Formula: C₃HBrN₂O₂S
MW: 209.02 g/mol
MDL Number: MFCD00005317
CAS Number: 3034-48-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty