Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isopropyldiphenylphosphine 98%

Supplier: Ambeed
Warning

A343809-250MG A343809-1G
77278-034EA 55 USD InStock
77278-034 77278-036
Isopropyldiphenylphosphine 98%
Isopropyldiphenylphosphine
Caution: for Research Use Only


Formula: C₁₅H₁₇P
MW: 228.27 g/mol
MDL Number: MFCD00015028
CAS Number: 6372-40-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty