Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2-Dichloroacetamide 95%

Supplier: Ambeed
Warning

A356167-100G A356167-10G A356167-500G A356167-25G
77251-606EA 29 USD InStock
77251-606 77251-608 77251-602 77251-604
2,2-Dichloroacetamide 95%
2,2-Dichloroacetamide
Caution: for Research Use Only


Formula: C₂H₃Cl₂NO
MW: 127.96 g/mol
MDL Number: MFCD00008015
CAS Number: 683-72-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty