Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dimethoxybenzoic acid 98%

Supplier: Ambeed
Warning

A385379-5G A385379-100G A385379-1G A385379-10G A385379-25G
77251-950EA 183 USD InStock
77251-950 77251-952 77251-958 77251-954 77251-956
2,5-Dimethoxybenzoic acid 98%
2,5-Dimethoxybenzoic acid
Caution: for Research Use Only


Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00002436
CAS Number: 2785-98-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty