Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylanisole 99%

Supplier: Ambeed
Danger
Synonyms: 2-Methoxytolueneo-Cresol methyll ether2-Methylanisole1-Methoxy-2-methylbenzene

A397991-5G A397991-25G
77278-706EA 6 USD InStock
77278-706 77278-708
2-Methylanisole 99%
2-Methylanisole
Caution: for Research Use Only


Formula: C₈H₁₀O
MW: 122.17 g/mol
MDL Number: MFCD00008373
CAS Number: 578-58-5
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty