Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Deoxyinosine 98%

Supplier: Ambeed
Warning
Synonyms: 9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol

A438356-1MG A438356-1G A438356-100MG
77264-954EA 6 USD InStock
77264-954 77264-958 77264-956
2'-Deoxyinosine 98%
2'-Deoxyinosine
Caution: for Research Use Only


Formula: C₁₀H₁₂N₄O₄
MW: 252.23 g/mol
MDL Number: MFCD00005762
CAS Number: 890-38-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty