Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylindoline 98%

Supplier: Ambeed
Warning

A460836-100G A460836-500G A460836-25G A460836-5G
77279-268EA 5 USD InStock
77279-268 77279-266 77279-264 77279-262
2-Methylindoline 98%
2-Methylindoline
Caution: for Research Use Only


Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00005706
CAS Number: 6872-06-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty